missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ADH4 Recombinant Rabbit Monoclonal Antibody (24GB5120)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556951
This item is not returnable.
View return policy
Description
ADH4 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene encodes class II alcohol dehydrogenase 4 pi subunit, which is a member of the alcohol dehydrogenase family. Members of this enzyme family metabolize a wide variety of substrates, including ethanol, retinol, other aliphatic alcohols, hydroxysteroids, and lipid peroxidation products. Class II alcohol dehydrogenase is a homodimer composed of 2 pi subunits. It exhibits a high activity for oxidation of long-chain aliphatic alcohols and aromatic alcohols and is less sensitive to pyrazole. This gene is localized to chromosome 4 in the cluster of alcohol dehydrogenase genes.
Specifications
| ADH4 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| ADH4 | |
| Ac1002; ADH-1; ADH2; ADH-2; Adh4; Alcohol dehydrogenase 4; alcohol dehydrogenase 4 (class II), pi polypeptide; alcohol dehydrogenase class II; Alcohol dehydrogenase class II pi chain; alcohol dehydrogenase II; aldehyde reductase; all-trans-retinol dehydrogenase [NAD(+)] ADH4; class II alcohol dehydrogenase, pi subunit; class II type ADH; epididymis secretory protein Li 4; HEL-S-4 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 127 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB5120 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P08319 | |
| ADH4 | |
| A synthesized peptide derived from human ADH4 (aa 300-380). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction