Learn More
ADH4 Mouse anti-Human, Unconjugated, Clone: 3D12E9, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67303-1-IG-20UL
 View more versions of this product
Product Code. 16829855
  • £135.00 / 20µL
Description
The ADH4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ADH4. This antibody recognizes human antigen. The ADH4 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
ADH4
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, ELISA
3D12E9
Unconjugated
20 μL
127
IgG1