Learn More
ADH1B Mouse anti-Human, Unconjugated, Clone: 1F1B4, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66939-1-IG-150UL
 View more versions of this product
Product Code. 16879245
  • £349.00 / 150µL
Description
The ADH1B antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ADH1B. This antibody recognizes human, mouse, rat, pig antigen. The ADH1B antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
ADH1B
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig
Western Blot, Immunohistochemistry, Immunofluorescence
1F1B4
Unconjugated
150 μL
125
IgG2a