missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ADGB Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557618
This item is not returnable.
View return policy
Description
Immunogen sequence: DGLNLEREIV SQTTATQEKS QEELPTTNNS VSKEIWLDFE DFCVCFQNIY IFHKPSSYCL NFQKSEFKFS EERVSYYLFV DSLKPIELLV CFSALVR Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 80%.
The function of this protein remains unknown.
Specifications
| ADGB | |
| Polyclonal | |
| Unconjugated | |
| ADGB | |
| ADGB; Androglobin; C6orf103; C9H6orf103; Calpain-7-like protein; CAPN16; CAPN7L | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79747 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N7X0 | |
| ADGB | |
| Recombinant protein corresponding to Human ADGB. Recombinant protein control fragment (Product #RP-102713). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction