missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADFP Mouse anti-Human, Clone: ADFP/1494, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB14487.100ug
This item is not returnable.
View return policy
Description
Mouse monoclonal antibody raised against partial recombinant human ADFP.
- High specificity
- High reactivity
Specifications
| ADFP | |
| Monoclonal | |
| Unconjugated | |
| Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| Q99541 | |
| ADFP | |
| Recombinant protein corresponding to amino acids 249-376 of human ADFP. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| ADFP/1494 | |
| Mouse monoclonal antibody raised against partial recombinant human ADFP. | |
| In 10 mM PBS (0.05% BSA, 0.05% sodium azide). | |
| ADRP/MGC10598 | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 123 | |
| Store at 4°C. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction