Learn More
3M™ Adflo™ Standard Lithium Ion Battery
Product Code. 15700018
Change view
Click to view available options
Battery Type: Lithium ion
Product Code.
15700018 Battery Type: Lithium ion
1 options
Product Code. 15700018 Supplier 3M™ Supplier No. 7000144538
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
3M™ Adflo™ Standard Lithium Ion Battery makes the Adflo™ Powered Air Respirator even more lightweight and balanced for greater comfort during the full shift.
  • Fully charges in 4.5 hours. Expected operating time is 7-9 hours with new particle filter, new fully-charged battery and a standard airflow
  • Batteries may be stored on the charger, fully charged, ready for immediate use
  • For use with the 3M™ Adflo™ Powered Air Respirator
TRUSTED_SUSTAINABILITY
Specifications
Battery Life 7 to 9 hr.
Battery Type Lithium-ion
Certifications/Compliance EN 12941
Product Type Standard Battery
Includes Particle filter (gas filter available as spare part)