missing translation for 'onlineSavingsMsg'
Learn More

Adezmapimod, MedChemExpress

Product Code. 16474806
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16474806 100 mg 100mg
16440127 50 mg 50mg
16413748 10 mg 10mg
16400966 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16474806 Supplier Medchem Express Supplier No. HY10256
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Adezmapimod (SB 203580) is a selective and ATP-competitive p38 MAPK inhibitor with IC50s of 50 nM and 500 nM for SAPK2a/p38 and SAPK2b/p38β2, respectively. Adezmapimod inhibits LCK, GSK3β and PKBα with IC50s of 100-500-fold higher than that for SAPK2a/p38. Adezmapimod does not disrupt JNK activity and is an autophagy and mitophagy activator.

Adezmapimod (SB 203580) is a selective and ATP-competitive p38 MAPK inhibitor with IC50s of 50 nM and 500 nM for SAPK2a/p38 and SAPK2b/p38β2, respectively. Adezmapimod inhibits LCK, GSK3β and PKBα with IC50s of 100-500-fold higher than that for SAPK2a/p38. Adezmapimod does not disrupt JNK activity and is an autophagy and mitophagy activator.

Specifications

Chemical Name or Material Adezmapimod
CAS 152121-47-6
Color White
Purity Grade Notes Research
Molecular Formula C21H16FN3OS
Quantity 100 mg
Synonym SB 203580 RWJ 64809
Solubility Information DMSO : 50 mg/mL (132.47 mM; Need ultrasonic)
SMILES O=S(C1=CC=C(C2=NC(C3=CC=C(F)C=C3)=C(C4=CC=NC=C4)N2)C=C1)C
Molecular Weight (g/mol) 377.43
Formula Weight 377.43
Percent Purity 96.59%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.