Learn More
Adenylosuccinate lyase Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15264-1-AP-150UL
 View more versions of this product
Product Code. 16824364
  • £349.00 / 150µL
Description
ADSL (adenylosuccinate lyase), also known as AMPS, ASL or ASASE, is a 484 amino acid protein that is involved in both purine biosynthesis and in the formation of adenosine monophosphate (AMP) from inosine monophosphate. Expressed ubiquitously, ADSL catalyzes two key reactions in AMP biosynthesis, namely the removal of a fumarate from succinylaminoimidazole carboxamide (SAICA) ribotide to give aminoimidazole carboxamide ribotide (AICA) and the subsequent removal of fumarate from adenylosuccinate to yield AMP. Defects in the gene encoding ADSL are the cause of adenylosuccinase deficiency (ADSL deficiency), an autosomal recessive disorder characterized by epilepsy, growth retardation and muscular wasting. Multiple isoforms of ADSL exist due to alternative splicing events.
Specifications
Specifications
Adenylosuccinate lyase
Polyclonal
Unconjugated
ADSL
Adenylosuccinase, adenylosuccinate lyase, ADSL, AMPS, ASASE, ASL
Rabbit
Antigen Affinity Chromatography
RUO
11564, 158, 315150
-20°C
Liquid
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.22 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P30566, P54822
ADSL
Adenylosuccinate lyase Fusion Protein Ag7332
150 μL
Primary
Human, Mouse, Rat