missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Adenylate Kinase 8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA521873
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji. Predicted reactivity: Bovine (83%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Enables AMP binding activity and nucleobase-containing compound kinase activity. Involved in nucleoside diphosphate phosphorylation and nucleoside triphosphate biosynthetic process. Located in 9+2 motile cilium.
Specifications
| Adenylate Kinase 8 | |
| Polyclonal | |
| Unconjugated | |
| Ak8 | |
| 1190002A17Rik; Adenylate kinase 8; Adenylate kinase isoenzyme 4, mitochondrial (ATP-AMP transphosphorylase); AK 8; AK8; ATP-AMP transphosphorylase 8; C9orf98; DDX31; HypB, AroK, ADK and rho factor domain containing protein RGD1303144; putative adenylate kinase-like protein C9orf98; putative adenylate kinase-like protein C9orf98 homolog; RGD1303144; RP11-143F18.1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 158067 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.62 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q96MA6 | |
| Ak8 | |
| Recombinant protein encompassing a sequence within the center region of human AK8. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction