missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Adenylate Kinase 8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104289
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CI098.
Enables AMP binding activity and nucleobase-containing compound kinase activity. Involved in nucleoside diphosphate phosphorylation and nucleoside triphosphate biosynthetic process. Located in 9+2 motile cilium.
Specifications
| Adenylate Kinase 8 | |
| Polyclonal | |
| Unconjugated | |
| Ak8 | |
| 1190002A17Rik; Adenylate kinase 8; Adenylate kinase isoenzyme 4, mitochondrial (ATP-AMP transphosphorylase); AK 8; AK8; ATP-AMP transphosphorylase 8; C9orf98; DDX31; HypB, AroK, ADK and rho factor domain containing protein RGD1303144; putative adenylate kinase-like protein C9orf98; putative adenylate kinase-like protein C9orf98 homolog; RGD1303144; RP11-143F18.1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 158067, 311833 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q68FP8, Q96MA6 | |
| Ak8 | |
| A synthesized peptide derived from human AK8(Accession Q96MA6), corresponding to amino acid residues Q342-R392. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction