missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Adenylate Kinase 8 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16577321
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16577321 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16577321 Supplier Invitrogen™ Supplier No. PA589416

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MDATIAPHRI PPEMPQYGEE NHIFELMQNM LEQLLIHQPE DPIPFMIQHL HRDNDNVPRI VILGPPASGK TTIAMWLCKH LNSSLLTLEN LILNEFSYTA TEARRLYLQR KTVPSALLVQ LIQERLAEED CIKQGWILDG IPETREQALR IQTLGITPRH VIVLSAPDTV LIERNLGKRI DPQTGEIYHT TFDWPPESEI; Positive Samples: Mouse brain, Mouse lung, Rat brain; Cellular Location: Cytoplasm, cytosol.

Enables AMP binding activity and nucleobase-containing compound kinase activity. Involved in nucleoside diphosphate phosphorylation and nucleoside triphosphate biosynthetic process. Located in 9+2 motile cilium.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Adenylate Kinase 8
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.22 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Ak8
Gene Accession No. Q32M07, Q68FP8
Gene Alias 1190002A17Rik; Adenylate kinase 8; Adenylate kinase isoenzyme 4, mitochondrial (ATP-AMP transphosphorylase); AK 8; AK8; ATP-AMP transphosphorylase 8; C9orf98; DDX31; HypB, AroK, ADK and rho factor domain containing protein RGD1303144; putative adenylate kinase-like protein C9orf98; putative adenylate kinase-like protein C9orf98 homolog; RGD1303144; RP11-143F18.1
Gene Symbols Ak8
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human AK8 (NP_6897851).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 311833, 68870
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.