missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Adenylate Kinase 5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563420
This item is not returnable.
View return policy
Description
Immunogen sequence: VVFLACANQR LKERLLKRAE QQGRPDDNVK ATQRRLMNFK QNAAPLVKYF QEKGLIMTFD ADRDEDEVFY DISMAVD Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 92%.
This gene encodes a member of the adenylate kinase family, which is involved in regulating the adenine nucleotide composition within a cell by catalyzing the reversible transfer of phosphate groups among adenine nucleotides. This member is related to the UMP/CMP kinase of several species. It is located in the cytosol and expressed exclusively in brain. Alternatively spliced transcript variants encoding distinct isoforms have been identified for this gene.
Specifications
| Adenylate Kinase 5 | |
| Polyclonal | |
| Unconjugated | |
| AK5 | |
| adenylate kinase 5; adenylate kinase 6; adenylate kinase isoenzyme 5; AK 5; AK5; AK6; ATP-AMP transphosphorylase 5; MGC33326; wu:fj63a06 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 26289 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y6K8 | |
| AK5 | |
| Recombinant protein corresponding to Human Adenylate Kinase 5. Recombinant protein control fragment (Product #RP-92480). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction