missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Adenylate Kinase 4 Recombinant Rabbit Monoclonal Antibody (23GB4755), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553843
This item is not returnable.
View return policy
Description
This gene encodes a member of the adenylate kinase family of enzymes. The encoded protein is localized to the mitochondrial matrix. Adenylate kinases regulate the adenine and guanine nucleotide compositions within a cell by catalyzing the reversible transfer of phosphate group among these nucleotides. Five isozymes of adenylate kinase have been identified in vertebrates. Expression of these isozymes is tissue-specific and developmentally regulated. A pseudogene for this gene has been located on chromosome 17. Three transcript variants encoding the same protein have been identified for this gene. Sequence alignment suggests that the gene defined by NM_013410, NM_203464, and NM_001005353 is located on chromosome 1.
Specifications
| Adenylate Kinase 4 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| AK4 | |
| adenylate kinase 3 alpha-like 1; Adenylate kinase 3-like; adenylate kinase 3-like {ECO:0000255; adenylate kinase 3-like 1; adenylate kinase 3-like 2; adenylate kinase 4; adenylate kinase 4, mitochondrial; adenylate kinase 4, mitochondrial {ECO:0000255; Adenylate kinase isoenzyme 4; adenylate kinase isoenzyme 4, mitochondrial; adenylate kinase isoenzyme 4, mitochondrial; adenylate kinase 4, mitochondrial; AK 4; AK 4 {ECO:0000255; AK3; Ak-3; Ak3b; Ak3l1; AK3L2; AK4; Ak-4; ATP-AMP transphosphorylase; D4Ertd274e; GTP:AMP phosphotransferase AK4; GTP:AMP phosphotransferase AK4 {ECO:0000255; GTP:AMP phosphotransferase AK4, mitochondrial; HAMAP-Rule:MF_03170}; mitochondrial adenylate kinase-3; nucleoside-triphosphate-adenylate kinase; RP4-686B20.1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 205 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB4755 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P27144 | |
| AK4 | |
| A synthesized peptide derived from human Adenylate kinase 4 (30-90AA). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction