missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenylate Kinase 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35229-100ul
This item is not returnable.
View return policy
Description
Adenylate Kinase 1 Polyclonal antibody specifically detects Adenylate Kinase 1 in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| Adenylate Kinase 1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| adenylate kinase 1, adenylate kinase isoenzyme 1, AK 1, ATP-AMP transphosphorylase 1, EC 2.7.4, EC 2.7.4.3, Myokinase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-194 of human Adenylate Kinase 1 (NP_000467.1).,, Sequence:, MEEKLKKTKIIFVVGGPGSGKGTQCEKIVQKYGYTHLSTGDLLRSEVSSGSARGKKLSEIMEKGQLVPLETVLDMLRDAMVAKVNTSKGFLIDGYPREVQQGEEFERRIGQPTLLLYVDAGPETMTQRLLKRGETSGRVDDNEETIKKRLETYYKATEPVIAFYEKRGIVRKVNAEGSVDSVFSQVCTHLDALK | |
| 100 μL | |
| metabolism, Signal Transduction | |
| 203 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction