missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenylate Kinase 1 Antibody (7E9), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59485AF350
This item is not returnable.
View return policy
Description
Adenylate Kinase 1 Monoclonal antibody specifically detects Adenylate Kinase 1 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| Adenylate Kinase 1 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| adenylate kinase 1, adenylate kinase isoenzyme 1, AK 1, ATP-AMP transphosphorylase 1, EC 2.7.4, EC 2.7.4.3, Myokinase | |
| Recombinant human Adenylate Kinase 1 (1-194aa) purified from E. coli | |
| 0.1 mL | |
| metabolism, Signal Transduction | |
| 203 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA, Immunofluorescence | |
| 7000000000 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction