Learn More
Abcam Adenylate Cyclase Activity Assay
Product Code. 32115688 Shop All Abcam Products
Change view
Click to view available options
Quantity:
1 x 100 Tests
Unit Size:
100 tests
Product Code. Quantity Unit Size
32115688 1 x 100 Tests 100 tests
1 options
Product Code. 32115688 Supplier Abcam Supplier No. ab3236081x100Tests
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Measure Adenylate Cyclase activity in 1h 15m in Cell/Tissue Extracts with Adenylate Cyclase Activity Assay AB323608. Sensitivity <0.02 mU. Use with Microplate.
Measure Adenylate Cyclase activity in 1h 15m in Cell/Tissue Extracts with Adenylate Cyclase Activity Assay AB323608. Sensitivity <0.02 mU. Use with Microplate.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 1 x 100 Tests