missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenylate Cyclase 9 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539560
This item is not returnable.
View return policy
Description
Adenylyl cyclases function to convert ATP to cyclic AMP in response to activation by a variety of hormones, neurotransmitters and other regulatory molecules. Cyclic AMP, in turn, activates several other target molecules (primarily cyclic AMP-dependent protein kinases) to control a broad range of diverse phenomena such as metabolism, gene transcription and memory. Classically, adenylyl cyclases respond to receptor-initiated signals, mediated by the Gs and Gi heterotrimeric G proteins. The binding of an agonist to a Gs-coupled receptor (i.e., alpha beta-adrenergic receptor) catalyzes the exchange of GDP (bound to G alpha s) for GTP, dissociation of GTP-G alpha sfrom G beta gamma and G alpha s-mediated activation of adenylyl cyclase. The most abundant cerebral adenylyl cyclases appears to be adenylyl cyclase IX. AC IX is confined to the gray matter and its expression is mainly neuronal, with its highest expression located at the hippocampus. ACIX is also expressed in heart, pancreas and thyrocytes. AC I and AC IX are regulated reciprocally by intracellular free Ca2+. The inhibition of AC IX by Ca2+ is blocked by the calcineurin inhibitors FK506 and cyclosporin A.
Specifications
| Adenylate Cyclase 9 | |
| Polyclonal | |
| Unconjugated | |
| ADCY9 | |
| Adenylate cyclase type 9, ATP pyrophosphate-lyase 9, Adenylate cyclase type IX, Adenylyl cyclase 9, ADCY9, KIAA0520 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 115 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O60503 | |
| ADCY9 | |
| A synthetic peptide derived from the N-terminal region of human ADCY9 | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction