missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Adenylate Cyclase 9 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16330505
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16330505 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16330505 Supplier Invitrogen™ Supplier No. PA583548

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: VADQLKGLKT YLISGQRAKE SRCSCAEALL SGFEVIDGSQ VSSGPRGQGT ASSGNVSDLA QTVKTFDNLK TCPSCGITFA PKSEAGAEGG APQNGCQD Highest antigen sequence indentity to the following orthologs: Rat - 94%, Mouse - 92%.

Adenylate cyclase is a membrane bound enzyme that catalyses the formation of cyclic AMP from ATP. It is regulated by a family of G protein-coupled receptors, protein kinases, and calcium. The type 9 adenylyl cyclase is a widely distributed adenylyl cyclase, and it is stimulated by beta-adrenergic receptor activation but is insensitive to forskolin, calcium, and somatostatin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Adenylate Cyclase 9
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ADCY9
Gene Accession No. O60503
Gene Alias AC9; ACIX; ACtp10; Adcy9; adenylate cyclase 9; adenylate cyclase type 9; Adenylate cyclase type IX; adenylyl cyclase 9; Adenylyl cyclase type 10; ATP pyrophosphate-lyase 9; AW125421; D16Wsu65e; KIAA0520; mKIAA0520; type IX ATP pyrophosphate-lyase
Gene Symbols ADCY9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ADCY9. Recombinant protein control fragment (Product #RP-98699).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.