missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Adenylate Cyclase 8 Polyclonal Antibody

Description
Immunogen sequence: DRRNSGATFT EGSWSPELPF DNIVGKQNTL AALTRNSINL LPNHLAQALH VQSGPEEINK RIEHTIDLRS GDKLRREHIK PFSLMF Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Adenylate cyclase is a membrane bound enzyme that catalyses the formation of cyclic AMP from ATP. The enzymatic activity is under the control of several hormones, and different polypeptides participate in the transduction of the signal from the receptor to the catalytic moiety. Stimulatory or inhibitory receptors (Rs and Ri) interact with G proteins (Gs and Gi) that exhibit GTPase activity and they modulate the activity of the catalytic subunit of the adenylyl cyclase.
Specifications
Specifications
| Antigen | Adenylate Cyclase 8 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Adcy8 |
| Gene Accession No. | P40145 |
| Gene Alias | AC8; ADCY3; ADCY8; adenylate cyclase 8; adenylate cyclase 8 (brain); adenylate cyclase type 8; Adenylate cyclase type VIII; adenylyl cyclase 8; adenylyl cyclase-8, brain; ATP pyrophosphate-lyase 8; AW060868; ca(2+)/calmodulin-activated adenylyl cyclase; epididymis secretory sperm binding protein Li 172mP; HBAC1; HEL-S-172mP; type VIII adenylyl cyclase |
| Gene Symbols | Adcy8 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?