missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenylate Cyclase 7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97909-50ul
This item is not returnable.
View return policy
Description
Adenylate Cyclase 7 Polyclonal antibody specifically detects Adenylate Cyclase 7 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Adenylate Cyclase 7 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| AC7, adenylate cyclase 7, adenylate cyclase type 7, Adenylate cyclase type VII, Adenylyl cyclase 7, ATP pyrophosphate-lyase 7, EC 4.6.1, EC 4.6.1.1, KIAA0037FLJ36387 | |
| Produced in rabbits immunized with E. coli-derived Human Adenylate Cyclase 7 fragment. | |
| 50 μg | |
| Lipid and Metabolism | |
| 113 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction