missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenylate Cyclase 7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-58945
This item is not returnable.
View return policy
Description
Adenylate Cyclase 7 Polyclonal specifically detects Adenylate Cyclase 7 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| Adenylate Cyclase 7 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1-4 ug/ml | |
| AC7, adenylate cyclase 7, adenylate cyclase type 7, Adenylate cyclase type VII, Adenylyl cyclase 7, ATP pyrophosphate-lyase 7, EC 4.6.1, EC 4.6.1.1, KIAA0037FLJ36387 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| ADCY7 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:PFAHLNHRESVSSGETHVPNGRRPKSVPQRHRRTPDRSMSPKGRSEDDSYDDEMLSAIEGLSSTRPCCSKSDDFYTFGSIFL | |
| 100 μL | |
| Lipid and Metabolism | |
| 113 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction