missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Adenylate Cyclase 5 Antibody (3E6), Novus Biologicals™

Product Code. 18343838 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18343838 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18343838 Supplier Novus Biologicals™ Supplier No. H00000111M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Adenylate Cyclase 5 Monoclonal antibody specifically detects Adenylate Cyclase 5 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Adenylate Cyclase 5
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 3E6
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunohistochemistry
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_899200
Gene Alias AC5, adenylate cyclase 5, adenylate cyclase type 5, Adenylate cyclase type V, Adenylyl cyclase 5, ATP pyrophosphate-lyase 5, EC 4.6.1.1
Host Species Mouse
Immunogen ADCY5 (NP_899200, 1152 a.a. ∽ 1261 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. ADFAMKLMDQMKYINEHSFNNFQMKIGLNIGPVVAGVIGARKPQYDIWGNTVNVASRMDSTGVPDRIQVTTDMYQVLAANTYQLECRGVVKVKGKGEMMTYFLNGGPPLS
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 111
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.