missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Adenylate Cyclase 3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA535382
This item is not returnable.
View return policy
Description
This antibody contains enough material to conduct 10 mini-Western blots.
This gene encodes adenylyl cyclase 3 which is a membrane-associated enzyme and catalyzes the formation of the secondary messenger cyclic adenosine monophosphate (cAMP). This protein appears to be widely expressed in various human tissues and may be involved in a number of physiological and pathophysiological metabolic processes.
Specifications
| Adenylate Cyclase 3 | |
| Polyclonal | |
| Unconjugated | |
| Adcy3 | |
| AC3; AC-III; ADCY3; adenylate cyclase 3; adenylate cyclase 8 (brain); adenylate cyclase type 3; Adenylate cyclase type III; adenylate cyclase, olfactive type; Adenylyl cyclase 3; adenylyl cyclase, type III; ATP pyrophosphate-lyase 3; KIAA0511; mKIAA0511; type III adenylyl cyclase | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 104111, 109, 64508 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 5mM sodium azide | |
| O60266, P21932, Q8VHH7 | |
| Adcy3 | |
| Synthetic peptide corresponding to the C-terminal region of rat ADCY3 conjugated to KLH. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction