Learn More
Invitrogen™ Adenylate Cyclase 3 Polyclonal Antibody
Chicken Polyclonal Antibody
Brand:  Invitrogen™ MA547338
 View more versions of this product
Product Code. 17896737
  • £1482.00 / 500µL
Description
Adenylate Cyclase 3 Polyclonal Antibody for Western Blot, ICC/IF
This gene encodes adenylyl cyclase 3 which is a membrane-associated enzyme and catalyzes the formation of the secondary messenger cyclic adenosine monophosphate (cAMP). This protein appears to be widely expressed in various human tissues and may be involved in a number of physiological and pathophysiological metabolic processes.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Adenylate Cyclase 3
Polyclonal
Unconjugated
Adcy3
AC3; AC-III; ADCY3; adenylate cyclase 3; adenylate cyclase 8 (brain); adenylate cyclase type 3; Adenylate cyclase type III; adenylate cyclase, olfactive type; Adenylyl cyclase 3; adenylyl cyclase, type III; ATP pyrophosphate-lyase 3; KIAA0511; mKIAA0511; type III adenylyl cyclase
Chicken
Affinity chromatography
RUO
104111, 64508
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot, Immunocytochemistry, Western Blot
1 mg/mL
PBS with 50% glycerol and 5mM sodium azide
P21932, Q8VHH7
Adcy3
C-terminal peptide of rat ACIII, PAAFPNGSSVTLPHQVVDNP with a Cys added to the N-terminus to allow coupling to KLH.
100 μL
Primary
Mouse, Rat
Antibody
IgY