missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenylate Cyclase 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £470.00
Specifications
| Antigen | Adenylate Cyclase 2 |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18481982
|
Novus Biologicals
NBP1-90281-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18230856
|
Novus Biologicals
NBP1-90281 |
0.1 mL |
£470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Adenylate Cyclase 2 Polyclonal specifically detects Adenylate Cyclase 2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Adenylate Cyclase 2 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| 3'-5'-cyclic AMP synthetase, AC2, adenylate cyclase 2 (brain), adenylate cyclase II, adenylate cyclase type 2, Adenylate cyclase type II, Adenylyl cyclase 2, ATP pyrophosphate-lyase 2, EC 4.6.1.1, FLJ45092, HBAC2, KIAA1060FLJ16822, MGC133314, type II adenylate cyclase | |
| ADCY2 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Polyclonal | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 108 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:HHRDSMTTENGKISTTDVPMGQHNFQNRTLRTKSQKKRFEEELNERMIQAIDGINAQKQWLKSEDIQRISLLFYNKVLEKEYRATA | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title