missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenylate Cyclase 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-15293
This item is not returnable.
View return policy
Description
Adenylate Cyclase 2 Polyclonal specifically detects Adenylate Cyclase 2 in Human samples. It is validated for Western Blot.
Specifications
| Adenylate Cyclase 2 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| 3'-5'-cyclic AMP synthetase, AC2, adenylate cyclase 2 (brain), adenylate cyclase II, adenylate cyclase type 2, Adenylate cyclase type II, Adenylyl cyclase 2, ATP pyrophosphate-lyase 2, EC 4.6.1.1, FLJ45092, HBAC2, KIAA1060FLJ16822, MGC133314, type II adenylate cyclase | |
| Rabbit | |
| 124 kDa | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 108 | |
| Human | |
| IgG |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q08462 | |
| ADCY2 | |
| Recombinant protein encompassing a sequence within the center region of human Adenylate cyclase 2. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Predicted cross-reactivity based on sequence identity: Rhesus Monkey (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction