missing translation for 'onlineSavingsMsg'
Learn More

Adenylate Cyclase 2 Antibody (1D4), Novus Biologicals™

Product Code. 18349977 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18349977 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18349977 Supplier Novus Biologicals Supplier No. H00000108M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Adenylate Cyclase 2 Monoclonal antibody specifically detects Adenylate Cyclase 2 in Human samples. It is validated for ELISA, Proximity Ligation Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen Adenylate Cyclase 2
Applications ELISA, Proximity Ligation Assay
Classification Monoclonal
Clone 1D4
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_065433
Gene Alias 3'-5'-cyclic AMP synthetase, AC2, adenylate cyclase 2 (brain), adenylate cyclase II, adenylate cyclase type 2, Adenylate cyclase type II, Adenylyl cyclase 2, ATP pyrophosphate-lyase 2, EC 4.6.1.1, FLJ45092, HBAC2, KIAA1060FLJ16822, MGC133314, type II adenylate cyclase
Host Species Mouse
Immunogen ADCY2 (NP_065433, 977 a.a. ~ 1086 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. GKLDAINKHSFNDFKLRVGINHGPVIAGVIGAQKPQYDIWGNTVNVASRMDSTGVLDKIQVTEETSLVLQTLGYTCTCRGIINVKGKGDLKTYFVNTEMSRSLSQSNVAS
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 108
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.