missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Adenovirus Native Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Concentration | 2.3 mg/mL |
| For Use With (Application) | ELISA |
| Formulation | Minimum Essential Media |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 30 kDa |
| Quantity | 1 mL |
| Storage Requirements | Store at -80°C. Avoid freeze/thaw cycles. |
| Gene Alias | Adenovirus |
| Research Category | Cell Biology, Stem Cell Markers, Virology Bacteria and Parasites |
| Protein | Adenovirus |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction