missing translation for 'onlineSavingsMsg'
Learn More

Adenovirus Antibody (8C4), DyLight 650, Novus Biologicals™

Product Code. 18421204 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.2 mL
Unit Size:
0.2mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18421204 0.2 mL 0.2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18421204 Supplier Novus Biologicals Supplier No. NB600413C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Adenovirus Monoclonal antibody specifically detects Adenovirus in Adenovirus, Virus samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunodiffusion, Sandwich ELISA, Lateral Flow, Neutralization
TRUSTED_SUSTAINABILITY

Specifications

Antigen Adenovirus
Applications ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunodiffusion, Sandwich ELISA
Classification Monoclonal
Clone 8C4
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias Adenovirus
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Human adenovirus type 1 and canine adenovirus type 1.
Purification Method Protein A purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Cell Biology, Stem Cell Markers, Virology Bacteria and Parasites
Primary or Secondary Primary
Target Species Adenovirus, Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.