missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenovirus-9 E4 Orf1 Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41088AF700
This item is not returnable.
View return policy
Description
Adenovirus-9 E4 Orf1 Polyclonal antibody specifically detects Adenovirus-9 E4 Orf1 in Adenovirus, Virus samples. It is validated for ELISA
Specifications
| Adenovirus-9 E4 Orf1 | |
| Polyclonal | |
| ELISA | |
| 13.6kDa protein, 14.5kDa protein, Control protein orf2, Control protein orf3, Control protein orf4, E4, E4 13.5kDa protein, E4 13.6kDa protein, E4 14.4kDa protein, E4 14.5kDa protein, E4 control protein 34 kDa, E4 control protein orf2, E4 control protein orf3, E4 control protein orf4, HuAdVD_gp17, ORF 2, Orf 3, Orf 4, ORF2, ORF3, ORF3 protein, ORF4, Putative E4 | |
| Antibody was raised against a 16 amino acid synthetic peptide near the carboxy terminus of the Ad-9 E4 Orf1. The immunogen is located within the last 50 amino acids of Adenovirus-9 E4 Orf1. Amino Acid Squence: FNHTDRFLYVREGHPV | |
| 0.1 mL | |
| Cancer | |
| 6386282 | |
| Store at 4C in the dark. | |
| IgG |
| ELISA, Immunohistochemistry, Immunofluorescence | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Adenovirus, Virus, Primate, Rabbit | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction