missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenosylhomocysteinase/AHCY Rabbit anti-Human, Mouse, Rat, Clone: 4E9H10, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-16147-100UL
This item is not returnable.
View return policy
Description
Adenosylhomocysteinase/AHCY Monoclonal antibody specifically detects Adenosylhomocysteinase/AHCY in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| Adenosylhomocysteinase/AHCY | |
| Monoclonal | |
| Unconjugated | |
| adenosylhomocysteinase, AdoHcyase, EC 3.3.1.1, S-adenosylhomocysteine hydrolase, S-adenosyl-L-homocysteine hydrolase, SAHHadoHcyase | |
| A synthetic peptide corresponding to a sequence within amino acids 300-400 of human Adenosylhomocysteinase/AHCY (P23526). | |
| 100 μg | |
| Amino Acids Drugs and other small molecules, Endocrinology, Signal Transduction | |
| 191 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 4E9H10 | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction