missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenosylhomocysteinase/AHCY Antibody (SR1809), Novus Biologicals™
Rabbit Monoclonal Antibody
£265.00 - £341.00
Specifications
| Antigen | Adenosylhomocysteinase/AHCY |
|---|---|
| Clone | SR1809 |
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18681045
|
Novus Biologicals
NBP3-22403-100ul |
100 μg |
£341.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18605384
|
Novus Biologicals
NBP3-22403-50ul |
50 μg |
£265.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Adenosylhomocysteinase/AHCY Monoclonal antibody specifically detects Adenosylhomocysteinase/AHCY in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Adenosylhomocysteinase/AHCY | |
| Western Blot 1:500-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| adenosylhomocysteinase, AdoHcyase, EC 3.3.1.1, S-adenosylhomocysteine hydrolase, S-adenosyl-L-homocysteine hydrolase, SAHHadoHcyase | |
| A synthesized peptide derived from Adenosylhomocysteinase/AHCY (Uniprot #: P23526) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1809 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Amino Acids Drugs and other small molecules, Endocrinology, Signal Transduction | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 191 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title