missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenosylhomocysteinase/AHCY Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20246
This item is not returnable.
View return policy
Description
Adenosylhomocysteinase/AHCY Polyclonal specifically detects Adenosylhomocysteinase/AHCY in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Adenosylhomocysteinase/AHCY | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| adenosylhomocysteinase, AdoHcyase, EC 3.3.1.1, S-adenosylhomocysteine hydrolase, S-adenosyl-L-homocysteine hydrolase, SAHHadoHcyase | |
| Rabbit | |
| 48 kDa | |
| 0.1 mL | |
| Primary | |
| Species reactivity based on sequence identity: Zebrafish (86%), Xenopus tropicalis (88%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 8.5mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| P23526 | |
| AHCY | |
| Recombinant protein encompassing a sequence within the center region of human AHCY. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 191 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction