missing translation for 'onlineSavingsMsg'
Learn More

Adenosine monophosphate, MedChemExpress

Product Code. 16472019
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16472019 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16472019 Supplier Medchem Express Supplier No. HYA0181
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Adenosine monophosphate is a key cellular metabolite regulating energy homeostasis and signal transduction.

Adenosine monophosphate is a key cellular metabolite regulating energy homeostasis and signal transduction.

Specifications

Chemical Name or Material Adenosine monophosphate
CAS 61-19-8
Color White
Purity Grade Notes Research
Molecular Formula C10H14N5O7P
Quantity 1 g
Synonym AMP
Solubility Information DMSO : 16.67 mg/mL (48.01 mM; Need ultrasonic) ∣H2O : 1.67 mg/mL (4.81 mM; Need ultrasonic)
SMILES O=P(O)(OC[C@@H]1[C@H]([C@H]([C@H](N2C=NC3=C2N=CN=C3N)O1)O)O)O
Molecular Weight (g/mol) 347.22
Formula Weight 347.22
Percent Purity 97.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.