missing translation for 'onlineSavingsMsg'
Learn More

Adenosine, MedChemExpress

Product Code. 16482099
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16482099 1 g 1g
16430838 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16482099 Supplier Medchem Express Supplier No. HYB0228
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Adenosine (Adenine riboside), a ubiquitous endogenous autacoid, acts through the enrollment of four G protein-coupled receptors: A1, A2A, A2B, and A3. Adenosine affects almost all aspects of cellular physiology, including neuronal activity, vascular function, platelet aggregation, and blood cell regulation.

Adenosine (Adenine riboside), a ubiquitous endogenous autacoid, acts through the enrollment of four G protein-coupled receptors: A1, A2A, A2B, and A3. Adenosine affects almost all aspects of cellular physiology, including neuronal activity, vascular function, platelet aggregation, and blood cell regulation.

Specifications

Chemical Name or Material Adenosine
CAS 58-61-7
Color White
Purity Grade Notes Research
Molecular Formula C10H13N5O4
Quantity 1 g
Synonym Adenine riboside D-Adenosine
Solubility Information DMSO : 33.33 mg/mL (124.72 mM; Need ultrasonic) ∣H2O : ≥ 6.67 mg/mL (24.96 mM)
SMILES NC1=C2C(N([C@H]3[C@H](O)[C@H](O)[C@@H](CO)O3)C=N2)=NC=N1
Molecular Weight (g/mol) 267.24
Formula Weight 267.24
Percent Purity 98.06%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.