missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenosine Deaminase/ADA Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-87404-25ul
This item is not returnable.
View return policy
Description
Adenosine Deaminase/ADA Polyclonal specifically detects Adenosine Deaminase/ADA in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.
Specifications
| Adenosine Deaminase/ADA | |
| Polyclonal | |
| Western Blot 0.04 - 0.4 ug/ml, Flow Cytometry Reported in the scientific literature (PMID: 25172498)., Immunohistochemistry 1:5000 - 1:10000, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:5000 - 1:10000, Knockdown Validated Reported in the scientific literature (PMID: 31978345). | |
| P00813 | |
| ADA | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:GDETIPGSSLLPGHVQAYQEAVKSGIHRTVHAGEVGSAEVVKEAVDILKTERLGHGYHTLEDQALYNRLRQENMHFEICPWSSYLTGAWKPDTEHAVIRLKNDQANYSLNTDDPLIFKSTLDTDYQMTKRDMGFTEEE | |
| Affinity Purified | |
| RUO | |
| 100 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| ADA1, adenine deaminase, Adenosine aminohydrolase, adenosine deaminase, EC 3.5.4.4 | |
| Rabbit | |
| 41 kDa | |
| 25 μL | |
| Primary | |
| Specificity of human Adenosine Deaminase/ADA antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction