missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenosine A3 R Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-26063
This item is not returnable.
View return policy
Description
Adenosine A3 R Polyclonal specifically detects Adenosine A3 R in Human, Rat, Bovine samples. It is validated for Peptide ELISA.
Specifications
| Adenosine A3 R | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| ADORA3 | |
| Peptide with sequence TASAICNYNAH corresponding to internal region according to NP_065734.5, NP_000668.1, NP_001075445.1. | |
| 0.1 mg | |
| GPCR, Growth and Development, Neuronal Cell Markers, Vision | |
| 140 | |
| Human, Rat, Bovine | |
| IgG |
| ELISA | |
| 0.5 mg/mL | |
| Peptide ELISA Detection limit 1:2000 | |
| A3AR, AD026, adenosine A3 receptor, adenosine receptor A3, bA552M11.5 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody is expected to recognize isoform 1 (NP_065734.5) and isoform 3 (NP_001075445.1). | |
| Store at -20C. Avoid freeze-thaw cycles. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu