missing translation for 'onlineSavingsMsg'
Learn More

Adenosine A2b R Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 4 publications

£173.00 - £456.00

Specifications

Antigen Adenosine A2b R
Concentration 1 mg/mL
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18633357
View Documents
Novus Biologicals
NBP2-41312-0.025mg
0.025 mg
£173.00
0.03mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18745663
View Documents
Novus Biologicals
NBP2-41312
0.1 mg
£456.00
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Adenosine A2b R Polyclonal specifically detects Adenosine A2b R in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

Adenosine A2b R
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human, Mouse, Rat
adenosine A2b receptor, adenosine receptor A2b, ADORA2
ADORA2B
IgG
Affinity Purified
41 kDa
1 mg/mL
Polyclonal
Rabbit
GPCR
PBS with 0.02% Sodium Azide
136
Antibody was raised against a 19 amino acid peptide near the carboxy terminus of human ADORA2B. The immunogen is located within the last 50 amino acids of ADORA2B. Amino Acid Squence: QADVKSGNGQAGVQPA
Primary
At least two isoforms of ADORA2B are known to exist; this antibody will detect both isoforms. ADORA2B antibody is predicted to not cross-react with ADORA2A.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.