missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenosine A2aR Antibody (Ig2838) - Chimeric - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£216.00 - £458.00
Specifications
| Antigen | Adenosine A2aR |
|---|---|
| Clone | Ig2838 |
| Concentration | 1 mg/mL |
| Applications | ELISA, Immunofluorescence, Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18604037
|
Novus Biologicals
NBP3-09003-0.2mg |
0.2 mg |
£458.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18619667
|
Novus Biologicals
NBP3-09003-0.025mg |
0.025 mg |
£216.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Adenosine A2aR Monoclonal antibody specifically detects Adenosine A2aR in Human samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence, NeutralizationSpecifications
| Adenosine A2aR | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| adenosine A2 receptor, adenosine A2a receptor, adenosine receptor A2a, adenosine receptor subtype A2a, ADORA 2, ADORA2, hA2aR, RDC8 | |
| Human recombinant A2-adenosine receptor. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Ig2838 | |
| ELISA, Immunofluorescence, Neutralization | |
| Unconjugated | |
| Mouse | |
| Apoptosis, GPCR, Immunology, Innate Immunity | |
| PBS | |
| 135 | |
| IgG1 κ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title