missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenosine A1R Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£292.00 - £488.00
Specifications
| Antigen | Adenosine A1R |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18256713
|
Novus Biologicals
NBP2-58443 |
100 μL |
£488.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18633348
|
Novus Biologicals
NBP2-58443-25ul |
25 μL |
£292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Adenosine A1R Polyclonal specifically detects Adenosine A1R in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| Adenosine A1R | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, GPCR, Growth and Development, Immunology, Innate Immunity, Neuronal Cell Markers, Signal Transduction, Vision | |
| adenosine A1 receptor, ADORA 1, RDC7adenosine receptor A1 | |
| ADORA1 | |
| IgG | |
| Affinity Purified |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| 134 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:NNLSAVERAWAANGSMGEPVIKCEFEKVISMEYMVYFN | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title