missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenosine A1R Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NB300-549FR
388.45 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Adenosine A1R Polyclonal antibody specifically detects Adenosine A1R in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Specifica
| Adenosine A1R | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| DyLight 680 | |
| adenosine A1 receptor, ADORA 1, RDC7adenosine receptor A1 | |
| Synthetic peptide corresponding to residues C(309) Q P K P P I D E D L P E E K A E D(326) of rat Adenosine Receptor A1. | |
| 0.1 mL | |
| Apoptosis, GPCR, Growth and Development, Immunology, Innate Immunity, Neuronal Cell Markers, Signal Transduction, Vision | |
| 134 | |
| Store at 4°C in the dark. | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto