missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenosine 5'-Triphosphate Disodium Salt, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | Adenosine-5'-triphosphate disodium salt |
| CAS | 987-65-5 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C10 H14 N5 O13 P3 . 2 Na |
| Quantity | 10 g |
| Synonym | Adenosine 5'-(tetrahydrogen triphosphate), sodium salt (1:2), Adenosine 5'-(tetrahydrogen triphosphate), disodium salt (8CI,9CI), ATP disodium, ATP disodium salt, Adenosine 5'-triphosphate disodium salt, Adenosine triphosphate disodium, Adenosine triphosphate disodium salt, Adetphos, Disodium ATP, Disodium adenosine 5'-triphosphate, Disodium adenosine triphosphate, Disodium adenosine-5'-triphosphoric acid, Disodium dihydrogen ATP, NSC 20268, Sodium ATP |
| InChI Formula | InChI=1S/C10H16N5O13P3.2Na/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(17)6(16)4(26-10)1-25-30(21,22)28-31(23,24)27-29(18,19)20;;/h2-4,6-7,10,16-17H,1H2,(H,21,22)(H,23,24)(H2,11,12,13)(H2,18,19,20);;/q;2*+1/p-2/t4-,6-,7-,10-;;/m1../s1 |
| SMILES | [Na+].[Na+].Nc1ncnc2c1ncn2[C@@H]3O[C@H](COP(=O)([O-])OP(=O)([O-])OP(=O)(O)O)[C@@H](O)[C@H]3O |
| IUPAC Name | disodium;[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] phosphono phosphate |
| Molecular Weight (g/mol) | 551.14 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction