missing translation for 'onlineSavingsMsg'
Learn More

Adenosine 5’-Triphosphate Disodium Salt, TRC

Product Code. 30313066
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 30313066

Brand: Toronto Research Chemicals TRCA28181050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Adenosine-5'-triphosphate disodium salt
CAS 987-65-5
Purity Grade Notes HPLC
Molecular Formula C10 H14 N5 O13 P3 . 2 Na
Quantity 50 mg
Synonym Adenosine 5'-(tetrahydrogen triphosphate), sodium salt (1:2), Adenosine 5'-(tetrahydrogen triphosphate), disodium salt (8CI,9CI), ATP disodium, ATP disodium salt, Adenosine 5'-triphosphate disodium salt, Adenosine triphosphate disodium, Adenosine triphosphate disodium salt, Adetphos, Disodium ATP, Disodium adenosine 5'-triphosphate, Disodium adenosine triphosphate, Disodium adenosine-5'-triphosphoric acid, Disodium dihydrogen ATP, NSC 20268, Sodium ATP
InChI Formula InChI=1S/C10H16N5O13P3.2Na/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(17)6(16)4(26-10)1-25-30(21,22)28-31(23,24)27-29(18,19)20;;/h2-4,6-7,10,16-17H,1H2,(H,21,22)(H,23,24)(H2,11,12,13)(H2,18,19,20);;/q;2*+1/p-2/t4-,6-,7-,10-;;/m1../s1
SMILES [Na+].[Na+].Nc1ncnc2c1ncn2[C@@H]3O[C@H](COP(=O)([O-])OP(=O)([O-])OP(=O)(O)O)[C@@H](O)[C@H]3O
IUPAC Name disodium;[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] phosphono phosphate
Molecular Weight (g/mol) 551.14
Formula Weight 550.9596
Percent Purity >95
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.