missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Adenosine-5'-triphosphate disodium salt hydrate, ultrapure, 98%

Product Code. p-7041805
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 15410137

Brand: Thermo Scientific Chemicals J60336.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 34369-07-8 | C10H14N5Na2O13P3 | 551.15 g/mol

Adenosine 5'-Triphosphate, is used as a p2 purinergicagonist. It is also used as a nucleotide, the 5'-triphosphate of deoxyadenosine activated precursor in DNA synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Adenosine 5′-Triphosphate, is used as a p2 purinergicagonist. It is also used as a nucleotide, the 5′-triphosphate of deoxyadenosine activated precursor in DNA synthesis.

Solubility
Soluble at 5% (w/v) in water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature -20°C. Keep away fom strong oxidizing agents. Stable under ordinary conditions. Light sensitive, Hygroscopic
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34369-07-8
Molecular Formula C10H14N5Na2O13P3
Molecular Weight (g/mol) 551.15
MDL Number MFCD00150755
InChI Key TTWYZDPBDWHJOR-WCYUCLFNNA-L
Synonym atp disodium salt hydrate, atp disodium salt, atp disodium hydrate, adenosine 5'-triphosphate disodium salt hydrate, atp disodium salt trihydrate, 56-65-5 non-sodium, 987-65-5 anhydrous, 5 inverted exclamation marka-atp-na2, adenosine 5'-triphosphate disodium sal
PubChem CID 131664345
IUPAC Name disodium;[[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl]oxy-hydroxyphosphoryl] phosphate;hydrate
SMILES [Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])(=O)OP([O-])(=O)OP(O)(O)=O)[C@@H](O)[C@H]3O)C2=NC=N1

Specifications

Color White
Quantity 25 g
Sensitivity Hygroscopic; Light sensitive
Merck Index 14,155
Solubility Information Soluble at 5% (w/v) in water
Formula Weight 605.19 (551.15 Anhydrous)
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Adenosine-5'-triphosphate disodium salt hydrate
Grade Ultrapure

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.