missing translation for 'onlineSavingsMsg'
Learn More

Adenosine 5'-Monophosphate Disodium Salt, TRC

Product Code. 17958134
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17958134 1 g 1g
17929104 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17958134 Supplier Toronto Research Chemicals Supplier No. TRCA2817901G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4578-31-8
Molecular Formula C10 H12 N5 O7 P . 2 Na
Molecular Weight (g/mol) 391.18
Synonym 5'-Adenylic acid sodium salt (1:2), 5'-Adenylic acid, disodium salt (7CI,8CI,9CI), 5'-AMP disodium salt, AMP disodium salt, Adenosine 5'-monophosphate disodium, Adenosine 5'-monophosphate disodium salt, Adenosine 5'-phosphate disodium salt, Adenylic acid disodium salt, Disodium 5'-AMP, Disodium 9-(β-D-ribofuranosyl)adenine 5'-monophosphate, Disodium AMP, Disodium adenosine 5'-phosphate, Disodium adenylate
IUPAC Name disodium;[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate
SMILES [Na+].[Na+].Nc1ncnc2c1ncn2[C@@H]3O[C@H](COP(=O)([O-])[O-])[C@@H](O)[C@H]3O

Specifications

Analyte or Component Names Adenosine 5'-Monophosphate Disodium Salt
Shipping Condition Room Temperature
InChI Formula InChI=1S/C10H14N5O7P.2Na/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(17)6(16)4(22-10)1-21-23(18,19)20;;/h2-4,6-7,10,16-17H,1H2,(H2,11,12,13)(H2,18,19,20);;/q;2*+1/p-2/t4-,6-,7-,10-;;/m1../s1
Solution Type Neat
Industry Type Forensics
Quantity 1 g
Formula Weight 391.027
Chemical Name or Material Adenosine 5'-Monophosphate Disodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.