missing translation for 'onlineSavingsMsg'
Learn More

Adenosine-5'-Monophosphate Disodium Salt, Thermo Scientific™

Product Code. 15845058 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Plastic bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15845058 25 g Plastic bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15845058 Supplier Thermo Scientific Supplier No. J10620.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 4578-31-8
Physical Form Powder
Quantity 25 g
Molecular Formula C10H12N5Na2O7P
MDL Number MFCD00065023
Beilstein 3586732
Synonym adenosine 5'-monophosphate disodium salt, sodium 2r,3s,4r,5r-5-6-amino-9h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl phosphate, disodium amp, amp disodium salt, disodium 5'-amp, adenosine phosphate disodium, 5'-amp disodium salt, 5'-adenylic acid, disodium salt, unii-t1wz11dsrn, adenylic acid disodium salt
InChI Key QGXLVXZRPRRCRP-IDIVVRGQSA-L
SMILES [Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1
IUPAC Name disodium [(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate
Molecular Weight (g/mol) 391.19
PubChem CID 20712
ChEBI CHEBI:81690
Formula Weight 391.18 (Anhydrous)
Packaging Plastic bottle
Chemical Name or Material Adenosine-5'-Monophosphate Disodium Salt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.