missing translation for 'onlineSavingsMsg'
Learn More

ADENOSINE-5'-DIPHOSPHATE, MP Biomedicals

Product Code. p-2861160 Shop All MP Biomedicals Products
Click to view available options
Quantity:
0.1 g
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 11480232

Brand: MP Biomedicals 02160053.1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 58-64-0
Molecular Formula C10H15N5O10P2
Molecular Weight (g/mol) 427.203
MDL Number MFCD00066473
InChI Key XTWYTFMLZFPYCI-KQYNXXCUSA-N
Synonym adenosine 5'-diphosphate, adenosine diphosphate, adp, adenosine-5'-diphosphate, adenosine pyrophosphate, adenosine 5'-trihydrogen diphosphate, adenosine-diphosphate, adenosine 5'-pyrophosphate, adp nucleotide, 5'-adenylphosphoric acid
PubChem CID 6022
ChEBI CHEBI:16761
IUPAC Name [(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphono hydrogen phosphate
SMILES C1=NC2=C(C(=N1)N)N=CN2C3C(C(C(O3)COP(=O)(O)OP(=O)(O)O)O)O

Specifications

Color White
Assay Percent Range >96%
Quantity 0.1 g
Formula Weight 427.2
Physical Form Solid
Chemical Name or Material Adenosine-5'-Diphosphate, Free acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.