missing translation for 'onlineSavingsMsg'
Learn More

Adenosine 5'-Diphosphate Disodium Salt, TRC

Product Code. 30321323
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30321323 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30321323 Supplier Toronto Research Chemicals Supplier No. TRCA2817031G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Adenosine 5'-Diphosphate Disodium Salt
CAS 16178-48-6
Molecular Formula C10 H13 N5 O10 P2 . 2 Na
Quantity 1 g
Synonym Adenosine 5'-(trihydrogen diphosphate), sodium salt (1:2), Adenosine 5'-(trihydrogen diphosphate), disodium salt (9CI), Adenosine-5'-(trihydrogen pyrophosphate), disodium salt (8CI), ADP disodium, ADP disodium salt, Adenosine-5'-diphosphate disodium salt, Disodium 5'-ADP, Disodium ADP, Disodium adenosine 5'-diphosphate, Trinosin S
InChI Formula InChI=1S/C10H15N5O10P2.2Na/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(17)6(16)4(24-10)1-23-27(21,22)25-26(18,19)20;;/h2-4,6-7,10,16-17H,1H2,(H,21,22)(H2,11,12,13)(H2,18,19,20);;/q;2*+1/p-2/t4-,6-,7-,10-;;/m1../s1
SMILES [Na+].[Na+].Nc1ncnc2c1ncn2[C@@H]3O[C@H](COP(=O)([O-])OP(=O)(O)[O-])[C@@H](O)[C@H]3O
IUPAC Name disodium;[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] hydrogen phosphate
Molecular Weight (g/mol) 471.16
Formula Weight 470.9933
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.