missing translation for 'onlineSavingsMsg'
Learn More

Adenosine-5'-Diphosphate Disodium Salt, MP Biomedicals™

Product Code. 11388329 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11388329 500 mg 500mg
11308339 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11388329 Supplier MP Biomedicals Supplier No. 02100056.5
  • ONLINE EXCLUSIVE PRICE
    £35.05
    £32.87 / 500mg
    Save £2.18
    6% Off
 Restricted Item
Estimated Shipment: 16-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Involved in energy storage and nucleic acid metabolism. Adenosine-5'-Diphosphate Disodium Salt, MP Biomedicals is an adenine nucleotide that converts into ATP by ATP synthases. ADP affects platelet activation through its interaction with ADP receptors.

ADP affects platelet activation through its interaction with ADP receptors P2Y1, P2Y12 and P2X1. Upon its conversion to adenosine by ecto-ADPases, platelet activation is inhibited via adenosine receptors.

Chemical Identifiers

CAS 16178-48-6
Molecular Formula C10H13N5Na2O10P2
Molecular Weight (g/mol) 471.17
MDL Number MFCD00150927
InChI Key ORKSTPSQHZNDSC-WCYUCLFNNA-L
Synonym adenosine 5'-diphosphate disodium salt, adenosine-5'-diphosphate disodium salt, sodium 2r,3s,4r,5r-5-6-amino-9h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl hydrogendiphosphate, adenosine 5'-diphosphate disodium salt hydrate, adenosine, 5'-diphosphoric acid, disodium salt, adenosine 5'-trihydrogen diphosphate , disodium salt, trinosin s hydrate, disodium hydrogen 2r,3s,4r,5r-5-6-aminopurin-9-yl-3,4-dihydroxyoxolan-2-yl methyl phosphonato oxyphosphonate, adenosine 5'-trihydrogen diphosphate , sodium salt 1:2, disodium adp hydrate
PubChem CID 85315
IUPAC Name disodium;[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] hydrogen phosphate
SMILES [Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1

Specifications

Quantity 500 mg
Optical Rotation [α]20/D -29.6±1° c = 0.5% in 0.5M Na2HPO4 (Lit.)
Formula Weight 471.2
Percent Purity ≥95%
Physical Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.