missing translation for 'onlineSavingsMsg'
Learn More

Adenosine-5'-Diphosphate Disodium Salt Dihydrate

Product Code. 15875048 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15875048 25 g Plastic bottle 25g
15865048 5 g Plastic bottle 5g
15885048 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15875048 Supplier Thermo Scientific Supplier No. J10490.14
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 16178-48-6 ∣ C10H13N5Na2O10P2 ∣ 471.17 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16178-48-6
Molecular Formula C10H13N5Na2O10P2
Molecular Weight (g/mol) 471.17
MDL Number MFCD00150927
InChI Key ORKSTPSQHZNDSC-WCYUCLFNNA-L
Synonym Adenosine-5 '-Diphosphate Disodium Salt Dihydrate, adenosine-5 '-diphosphate disodium salt, sodium 2 r,3 s,4 r,5 r-5-6-amino-9 h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl hydrogendiphosphate, adenosine 5 '-diphosphate disodium salt hydrate, adenosine, 5 '-diphosphoric acid, disodium salt, adenosine 5'-trihydrogen diphosphate , disodium salt, trinosin s hydrate, disodium hydrogen 2r,3s,4r,5r-5-6-aminopurin-9-yl-3,4-dihydroxyoxolan-2-yl methyl phosphonato oxyphosphonate, adenosine 5'-trihydrogen diphosphate , sodium salt 1:2, disodium adp hydrate
PubChem CID 85315
SMILES [Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1

Specifications

Physical Form Powder
Quantity 25 g
Beilstein 3645656
IUPAC Name disodium;[[(2 R,3 S,4 R,5 R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] hydrogen phosphate
Formula Weight 507.2
Grade Ungraded
Packaging Plastic bottle
Chemical Name or Material Adenosine-5 '-Diphosphate Disodium Salt Dihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.